AMBH2D6FB064 Ambeed, Inc.

5-tert-Butyl 4-ethyl 3-methyl-6,7-dihydro-3H-imidazo[4,5-c]pyridine-4,5(4H)-dicarboxylate

  • MDL number MFCD22393526
  • Cas Number : 1380679-97-9
  • Molecular Weight : 309.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1380679-97-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1380679-97-9.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart