AMBH2D6FB0A3 Ambeed, Inc.

(1-Isopropyl-3-(trifluoromethyl)-1H-pyrazol-4-yl)boronic acid

  • MDL number MFCD25953898
  • Cas Number : 2170881-31-7
  • Molecular Weight : 221.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Shipped in cold pack
Country of Origin CN
Product Link https://www.ambeed.com/products/2170881-31-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2170881-31-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart