AMBH2D6FB0C7 Ambeed, Inc.

Methyl 2-(bromomethyl)-5-nitrobenzoate

  • MDL number MFCD04114313
  • Cas Number : 90725-68-1
  • Molecular Weight : 274.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Flash Point 194.1°C
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 397.3°C at 760 mmHg
Product Link https://www.ambeed.com/products/90725-68-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/90725-68-1.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart