AMBH2D6FB132 Ambeed, Inc.

Doxycycline hydrochloride

  • MDL number MFCD03427564
  • Cas Number : 10592-13-9
  • Molecular Weight : 480.90

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/10592-13-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/10592-13-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart