AMBH2D6FB19F Ambeed, Inc.

(3-(Benzylamino)oxetan-3-yl)methanol

  • MDL number MFCD18762004
  • Cas Number : 1195684-52-6
  • Molecular Weight : 193.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 153.6°C
Storage temp. Keep in dark place,Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 330.4°32.0°C at 760 mmHg
Product Link https://www.ambeed.com/products/1195684-52-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1195684-52-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart