AMBH2D6FB22A Ambeed, Inc.

5,6-Dibromo-1,2-dihydroacenaphthylene

  • MDL number MFCD00191989
  • Cas Number : 19190-91-1
  • Molecular Weight : 312.00

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 95
Flash Point 216.8°C
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 383°C at 760 mmHg
Product Link https://www.ambeed.com/products/19190-91-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/19190-91-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart