AMBH2D6FB239 Ambeed, Inc.

2-Bromo-5H-pyrrolo[2,3-b]pyrazine-7-carboxylic acid

  • MDL number MFCD26793713
  • Cas Number : 1422772-78-8
  • Molecular Weight : 242.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1422772-78-8.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/1422772-78-8.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart