AMBH2D6FB260 Ambeed, Inc.

8-Methoxypyrido[3,4-d]pyrimidin-4(3H)-one

  • MDL number MFCD21648261
  • Cas Number : 1260178-71-9
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1260178-71-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1260178-71-9.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart