AMBH2D6FB2E4 Ambeed, Inc.

2-Chloro-6,7-dihydro-5H-pyrrolo[2,3-d]pyrimidine

  • MDL number MFCD22069613
  • Cas Number : 1367986-07-9
  • Molecular Weight : 155.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 177.8
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 370.5�0�t 760 mmHg
Product Link https://www.ambeed.com/products/1367986-07-9.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/1367986-07-9.html
pictograms GHS07
hstatements H302-H319
Availability details for
Quantity
  • Availability
  • Add to Cart