AMBH2D6FB2FA Ambeed, Inc.

EW 7197

  • MDL number N/A
  • Cas Number : 1352608-82-2
  • Molecular Weight : 399.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-((4-([1,2,4]Triazolo[1,5-a]pyridin-6-yl)-5-(6-methylpyridin-2-yl)-1H-imidazol-2-yl)methyl)-2-fluoroaniline
Purity 98
Storage temp. Keep in dark place,Sealed in dry,Store in freezer, under -20
Physical Form solid
Shipping Temperature Shipped in cold pack
Country of Origin CN
Product Link https://www.ambeed.com/products/1352608-82-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1352608-82-2.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart