AMBH2D6FB31F Ambeed, Inc.

1,3,6,8-Tetrabromocarbazole

  • MDL number MFCD00218272
  • Cas Number : 55119-09-0
  • Molecular Weight : 482.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 280.6
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 540.4�t 760 mmHg
Product Link https://www.ambeed.com/products/55119-09-0.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/55119-09-0.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart