AMBH2D6FB3BC Ambeed, Inc.

1-(3-Nitropyridin-2-yl)ethanone

  • MDL number MFCD18904525
  • Cas Number : 194278-44-9
  • Molecular Weight : 166.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/194278-44-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/194278-44-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart