AMBH2D6FB3F0 Ambeed, Inc.

5-Chloro-7-methyl-1h-indole-2-carboxylic acid

  • MDL number MFCD06737780
  • Cas Number : 15936-74-0
  • Molecular Weight : 209.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/15936-74-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/15936-74-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart