AMBH2D6FBCC9 Ambeed, Inc.

1-(3-Bromo-2-fluorophenyl)ethanone

  • MDL number MFCD09264507
  • Cas Number : 161957-61-5
  • Molecular Weight : 217.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8°C
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/161957-61-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/161957-61-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart