AMBH2D6FBDC7 Ambeed, Inc.

7-Iodo-4-chloroquinoline

  • MDL number MFCD00511002
  • Cas Number : 22200-50-6
  • Molecular Weight : 289.50

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/22200-50-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/22200-50-6.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart