AMBH2D6FBE00 Ambeed, Inc.

2-(4-Fluorophenyl)-4,6-diphenyl-1,3,5-triazine

  • MDL number MFCD28964710
  • Cas Number : 203450-08-2
  • Molecular Weight : 327.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/203450-08-2.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/203450-08-2.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart