AMBH2D6FBF0F Ambeed, Inc.

2,7-Dibromo-9-phenyl-9H-carbazole

  • MDL number MFCD28147705
  • Cas Number : 444796-09-2
  • Molecular Weight : 401.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 505.3�t 760 mmHg
Iupac Name 2,7-dibromo-9-phenyl-9H-carbazole
Inchi Code 1S/C18H11Br2N/c19-12-6-8-15-16-9-7-13(20)11-18(16)21(17(15)10-12)14-4-2-1-3-5-14/h1-11H
Inchi Key MDXCDMSVFQIDGN-UHFFFAOYSA-N

Description

Purity 95%
Flash Point 259.4
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 505.3�t 760 mmHg
Product Link https://www.ambeed.com/products/444796-09-2.html

Safety Information

signalword Warning
pcodes P273-P305+P351+P338
MSDS https://www.ambeed.com/products/444796-09-2.html
pictograms GHS07
hstatements H302-H315-H319-H412
Availability details for
Quantity
  • Availability
  • Add to Cart