AMBH2D6FBF1B Ambeed, Inc.

1-Fluoro-3-(2-methoxyethoxy)-2-nitrobenzene

  • MDL number MFCD13689264
  • Cas Number : 131055-85-1
  • Molecular Weight : 215.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/131055-85-1.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/131055-85-1.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart