AMBH2D6FBF33 Ambeed, Inc.

2,4-Dichloro-8-methoxyquinoline

  • MDL number MFCD07644554
  • Cas Number : 32608-29-0
  • Molecular Weight : 228.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 327.9�t 760 mmHg
Product Link https://www.ambeed.com/products/32608-29-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/32608-29-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart