AMBH2D6FBF4D Ambeed, Inc.

(R)-3-Aminobutanenitrile hydrochloride

  • MDL number MFCD18651597
  • Cas Number : 1073666-55-3
  • Molecular Weight : 120.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1073666-55-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/1073666-55-3.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart