AMBH2D6FBF79 Ambeed, Inc.

(R)-1-(5-Fluoropyridin-2-yl)ethanamine hydrochloride

  • MDL number MFCD12759620
  • Cas Number : 1202070-39-0
  • Molecular Weight : 176.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 95
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1202070-39-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1202070-39-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart