AMBH2D6FBFA4 Ambeed, Inc.

tert-Butyl 7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydrobenzo[f][1,4]oxazepine-4(5H)-carboxylate

  • MDL number MFCD14582651
  • Cas Number : 1256784-52-7
  • Molecular Weight : 375.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form Colorless to White to Yellow Solid or Liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1256784-52-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1256784-52-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart