AMBH2D6FBFA9 Ambeed, Inc.

Methyl 4-bromo-2-methyl-1H-indole-6-carboxylate

  • MDL number MFCD18380825
  • Cas Number : 1260383-49-0
  • Molecular Weight : 268.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1260383-49-0.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/1260383-49-0.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart