AMBH2D6FBFCA Ambeed, Inc.

3,4-Dihydro-2H-pyrano[2,3-c]pyridine-6-carbaldehyde

  • MDL number MFCD18802894
  • Cas Number : 527681-61-4
  • Molecular Weight : 163.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58-60
Purity 95
Flash Point 127.8
Storage temp. Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Shipped in cold pack
Country of Origin CN
Boiling point 287.8�0�t 760 mmHg
Product Link https://www.ambeed.com/products/527681-61-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/527681-61-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart