AMBH2D6FC7E8 Ambeed, Inc.

(R)-2-Amino-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl)propanoic acid

  • MDL number MFCD00063056
  • Cas Number : 51-49-0
  • Molecular Weight : 776.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/51-49-0.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/51-49-0.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart