AMBH2D6FC7EA Ambeed, Inc.

(R)-N-(2,6-Dimethylphenyl)-1-propylpiperidine-2-carboxamide

  • MDL number MFCD06412576
  • Cas Number : 98717-16-9
  • Molecular Weight : 274.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/98717-16-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/98717-16-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart