AMBH2D6FC819 Ambeed, Inc.

(2,4,6-Trichlorophenyl)hydrazine

  • MDL number MFCD00025090
  • Cas Number : 5329-12-4
  • Molecular Weight : 211.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/5329-12-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/5329-12-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart