AMBH2D6FC850 Ambeed, Inc.

2-Amino-2-cyclohexylacetic acid

  • MDL number MFCD05662746
  • Cas Number : 5664-29-9
  • Molecular Weight : 157.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 256
Purity 97
Flash Point 131
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 292.8�t 760 mmHg
Product Link https://www.ambeed.com/products/5664-29-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/5664-29-9.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart