AMBH2D6FC908 Ambeed, Inc.

6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

  • MDL number MFCD07368852
  • Cas Number : 642494-36-8
  • Molecular Weight : 243.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/642494-36-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/642494-36-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart