AMBH2D6FCA51 Ambeed, Inc.

4-Bromo-2,5-dimethylphenol

  • MDL number MFCD00029746
  • Cas Number : 85223-93-4
  • Molecular Weight : 201.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 110.5
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 259°C at 760 mmHg
Product Link https://www.ambeed.com/products/85223-93-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/85223-93-4.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart