AMBH2D6FCAB7 Ambeed, Inc.

Methyl 3-iodo-4-methylbenzoate

  • MDL number MFCD00230583
  • Cas Number : 90347-66-3
  • Molecular Weight : 276.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 137
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form Form: Crystal - Lump - Liquid / Colour: Colorless - White - Yellow
Shipping Temperature Normal
Country of Origin CN
Boiling point 302.9�t 760 mmHg
Product Link https://www.ambeed.com/products/90347-66-3.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/90347-66-3.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart