AMBH2D6FCABD Ambeed, Inc.

7-Methyl-1,2,3,4-tetrahydroquinoline hydrochloride

  • MDL number MFCD09026784
  • Cas Number : 90874-58-1
  • Molecular Weight : 183.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/90874-58-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/90874-58-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart