AMBH2D6FCB26 Ambeed, Inc.

5-Ethynylisoquinoline

  • MDL number MFCD07644619
  • Cas Number : 1203579-37-6
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 298.1�0�t 760 mmHg
Product Link https://www.ambeed.com/products/1203579-37-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1203579-37-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart