AMBH2D6FCB41 Ambeed, Inc.

(2-Chlorophenyl)hydrazine hydrochloride

  • MDL number MFCD00012928
  • Cas Number : 41052-75-9
  • Molecular Weight : 179.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200-203
Purity 97
Flash Point 106.2
Storage temp. Keep in dark place,Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 252.1�t 760 mmHg
Product Link https://www.ambeed.com/products/41052-75-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/41052-75-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart