AMBH2D6FCB93 Ambeed, Inc.

(6-Methoxypyridin-2-yl)boronic acid

  • MDL number MFCD03093332
  • Cas Number : 372963-51-4
  • Molecular Weight : 152.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 150.8
Storage temp. Inert atmosphere,Store in freezer, under -20°C
Physical Form solid
Shipping Temperature Shipped in cold pack
Country of Origin CN
Boiling point 325.7±52.0°C at 760 mmHg
Product Link https://www.ambeed.com/products/372963-51-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/372963-51-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart