AMBH2D6FCB98 Ambeed, Inc.

2,4-Dichloro-6-methoxyquinoline

  • MDL number MFCD03285967
  • Cas Number : 70049-46-6
  • Molecular Weight : 228.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/70049-46-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/70049-46-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart