AMBH2D6FCBDA Ambeed, Inc.

2-Mercapto-4,6-dimethylpyrimidine

  • MDL number MFCD00006079
  • Cas Number : 22325-27-5
  • Molecular Weight : 140.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 213-216
Iupac Name 4,6-dimethyl-2(1H)-pyrimidinethione
Inchi Code 1S/C6H8N2S/c1-4-3-5(2)8-6(9)7-4/h3H,1H2,2H3,(H2,7,8,9)
Inchi Key JHIYVFQMMZLRNU-UHFFFAOYSA-N

Description

Melting Point 213-216
Purity 98%15%2
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/22325-27-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/22325-27-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart