AMBH2D6FD464 Ambeed, Inc.

(5-Chloro-2-fluoropyridin-3-yl)boronic acid

  • MDL number MFCD04972408
  • Cas Number : 937595-70-5
  • Molecular Weight : 175.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 346.6°C at 760 mmHg
Product Link https://www.ambeed.com/products/937595-70-5.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/937595-70-5.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart