AMBH2D6FD625 Ambeed, Inc.

2-(Benzylthio)quinoline

  • MDL number MFCD00093160
  • Cas Number : 116249-87-7
  • Molecular Weight : 251.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/116249-87-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/116249-87-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart