AMBH2D6FD645 Ambeed, Inc.

3-(4-Fluorophenyl)-1H-indole

  • MDL number MFCD03840861
  • Cas Number : 101125-32-0
  • Molecular Weight : 211.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/101125-32-0.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/101125-32-0.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart