AMBH2D6FD700 Ambeed, Inc.

2-Amino-3-mercaptopropanoic acid hydrochloride

  • MDL number MFCD00064552
  • Cas Number : 10318-18-0
  • Molecular Weight : 157.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 109-112
Iupac Name hydrogen chloride compound with cysteine (1:1)
Inchi Code 1S/C3H7NO2S.ClH/c4-2(1-7)3(5)6;/h2,7H,1,4H2,(H,5,6);1H
Inchi Key IFQSXNOEEPCSLW-UHFFFAOYSA-N

Description

Melting Point 109-112
Synonym 2-Amino-3-mercaptopropanoic acid hydrochloride
Purity 95%
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/10318-18-0.html

Safety Information

signalword Warning
pcodes P261-P264-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P321-P332+P313-P337+P313-P362-P403+P233-P405-P501
MSDS https://www.ambeed.com/products/10318-18-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart