AMBH2D6FD8F7 Ambeed, Inc.

2,5-Dibromo-4-methylimidazole

  • MDL number MFCD02179519
  • Cas Number : 219814-29-6
  • Molecular Weight : 239.90

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/219814-29-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/219814-29-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart