AMBH2D6FD9FB Ambeed, Inc.

Boc-trans-4-Tosyloxy-L-proline methyl ester

  • MDL number MFCD02179383
  • Cas Number : 88043-21-4
  • Molecular Weight : 399.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/88043-21-4.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/88043-21-4.html
pictograms GHS07
hstatements H319
Availability details for
Quantity
  • Availability
  • Add to Cart