AMBH2D6FDA22 Ambeed, Inc.

3-(Trifluoromethyl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD07375188
  • Cas Number : 1001020-14-9
  • Molecular Weight : 164.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1001020-14-9.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1001020-14-9.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart