AMBH2D6FDA9A Ambeed, Inc.

1,2,3,4-Tetrahydroquinolin-3-amine

  • MDL number MFCD01318268
  • Cas Number : 40615-02-9
  • Molecular Weight : 148.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 354.4�t 760 mmHg
Product Link https://www.ambeed.com/products/40615-02-9.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/40615-02-9.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart