AMBH2D6FDABA Ambeed, Inc.

4-Bromo-5-methyl-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole

  • MDL number MFCD19443358
  • Cas Number : 1926172-50-0
  • Molecular Weight : 295.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1926172-50-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1926172-50-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart