AMBH2D6FDC68 Ambeed, Inc.

8-Chloro-7-iodoimidazo[1,2-a]pyridine

  • MDL number MFCD25542237
  • Cas Number : 1447607-66-0
  • Molecular Weight : 278.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1447607-66-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1447607-66-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart