AMBH2D6FDC79 Ambeed, Inc.

2-Amino-5-bromo-3-(hydroxymethyl)pyridine hydrobromide

  • MDL number MFCD08272105
  • Cas Number : 443956-55-6
  • Molecular Weight : 283.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/443956-55-6.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/443956-55-6.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart