AMBH2D6FDC82 Ambeed, Inc.

(R)-2-Aminopropanamide

  • MDL number MFCD00672501
  • Cas Number : 35320-22-0
  • Molecular Weight : 88.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/35320-22-0.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/35320-22-0.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart