AMBH2D6FDC9D Ambeed, Inc.

(R)-2-Amino-2-(3-fluoro-2-methylphenyl)ethanol hydrochloride

  • MDL number MFCD24429021
  • Cas Number : 1956437-40-3
  • Molecular Weight : 205.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 124.2
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 281.8��t 760 mmHg
Product Link https://www.ambeed.com/products/1956437-40-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1956437-40-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart